(5-Bromo-2,3,4-trifluorophenyl)(cyclobutyl)methanone ≥97%

Supplier: AOBChem USA

Warning

377973-5G 377973-1G 377973-25G 377973-500MG
77628-940EA 1484.23 USD
77628-940 77638-468 77659-044 77650-486
(5-Bromo-2,3,4-trifluorophenyl)(cyclobutyl)methanone ≥97%
(5-Bromo-2,3,4-trifluorophenyl)(cyclobutyl)methanone

Precaución: For Research Use Only

Formula: C₁₁H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD35197875
CAS Number: 3002461-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR