(4,5-Dichloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

37642-5G 37642-1G 37642-25G 37642-500MG
77633-006EA 1115 USD
77633-006 77638-466 77655-490 77650-484
(4,5-Dichloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥95%
(4,5-Dichloro-2-fluoro-3-methylphenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 225.111 g/mol
MW: C₈H₇Cl₂FS
Storage Temperature: Ambient
MDL Number: MFCD32902608
CAS Number: 2586127-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR