(Z)-1-(2,6-Dichloro-3-ethoxyphenyl)but-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

375959-5G 375959-25G 375959-1G 375959-500MG
77628-938EA 1484.23 USD
77628-938 77659-040 77642-474 77650-482
(Z)-1-(2,6-Dichloro-3-ethoxyphenyl)but-2-en-1-ol ≥97%
(Z)-1-(2,6-Dichloro-3-ethoxyphenyl)but-2-en-1-ol

Precaución: For Research Use Only

Formula: 261.144 g/mol
MW: C₁₂H₁₄Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD35197109
CAS Number: 3002983-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR