3,4,5-Tribromoaniline ≥95%

Supplier: AOBChem USA

Warning

37590-5G 37590-25G 37590-1G 37590-500MG
77633-004EA 799 USD
77633-004 77655-486 77642-472 77646-484
3,4,5-Tribromoaniline ≥95%
3,4,5-Tribromoaniline

Precaución: For Research Use Only

Formula: 329.815 g/mol
MW: C₆H₄Br₃N
Storage Temperature: Ambient
MDL Number: MFCD02657490
CAS Number: 609-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR