1-(2,6-Difluoro-3-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
375665-5G
375665-1G
375665-25G
375665-500MG
77628-934EA
1484.23
USD
77628-934
77638-462
77659-038
77650-478
1-(2,6-Difluoro-3-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%
1-(2,6-Difluoro-3-(trifluoromethyl)phenyl)prop-2-en-1-ol
Precaución: For Research Use Only
Formula:
238.154 g/mol MW: C₁₀H₇F₅O Storage Temperature: Ambient |
MDL Number:
MFCD35197006 CAS Number: 149946-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...