Ethyl 2-(2,6-difluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

375664-5G 375664-25G 375664-1G 375664-500MG
77628-932EA 1484.23 USD
77628-932 77635-738 77642-468 77650-476
Ethyl 2-(2,6-difluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,6-difluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 282.163 g/mol
MW: C₁₁H₇F₅O₃
Storage Temperature: Ambient
MDL Number: MFCD35197005
CAS Number: 3002433-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR