(2,6-Difluoro-3-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

375659-5G 375659-25G 375659-1G 375659-500MG
77633-002EA 1441 USD
77633-002 77655-482 77642-466 77650-474
(2,6-Difluoro-3-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
(2,6-Difluoro-3-(trifluoromethyl)phenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 288.213 g/mol
MW: C₁₄H₉F₅O
Storage Temperature: Ambient
MDL Number: MFCD35197000
CAS Number: 3002516-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR