2-(2,6-Difluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

375653-5G 375653-25G 375653-1G 375653-500MG
77628-930EA 1484.23 USD
77628-930 77655-480 77642-464 77650-472
2-(2,6-Difluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(2,6-Difluoro-3-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 240.17 g/mol
MW: C₁₀H₉F₅O
Storage Temperature: Ambient
MDL Number: MFCD35196994
CAS Number: 3002468-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR