Cyclopropyl(2,6-difluoro-3-(trifluoromethyl)phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

375639-5G 375639-25G 375639-1G 375639-500MG
77628-924EA 1491.44 USD
77628-924 77635-734 77642-462 77646-480
Cyclopropyl(2,6-difluoro-3-(trifluoromethyl)phenyl)methanone ≥95%
Cyclopropyl(2,6-difluoro-3-(trifluoromethyl)phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₁H₇F₅O
Storage Temperature: Ambient
MDL Number: MFCD35196980
CAS Number: 3002433-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR