Methyl 2-(2,6-difluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

375635-5G 375635-25G 375635-500MG 375635-1G
77628-922EA 1484.23 USD
77628-922 77635-732 77652-734 77642-460
Methyl 2-(2,6-difluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Methyl 2-(2,6-difluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 268.137 g/mol
MW: C₁₀H₅F₅O₃
Storage Temperature: Ambient
MDL Number: MFCD35196976
CAS Number: 3002426-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR