(2,6-Difluoro-3-(trifluoromethyl)phenyl)trimethylsilane ≥95%
Supplier: AOBChem USA
|
Warning
|
375628-5G
375628-1G
375628-25G
375628-500MG
77633-000EA
1441
USD
77633-000
77638-454
77659-032
77650-468
(2,6-Difluoro-3-(trifluoromethyl)phenyl)trimethylsilane ≥95%
(2,6-Difluoro-3-(trifluoromethyl)phenyl)trimethylsilane
Precaución: For Research Use Only
Formula:
254.272 g/mol MW: C₁₀H₁₁F₅Si Storage Temperature: Ambient |
MDL Number:
MFCD35196969 CAS Number: 3002538-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...