1-(Cyclopropylethynyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

375587-5G 375587-1G 375587-25G 375587-500MG
77632-998EA 1357 USD
77632-998 77638-450 77655-478 77646-476
1-(Cyclopropylethynyl)cyclohexanol ≥95%
1-(Cyclopropylethynyl)cyclohexanol

Precaución: For Research Use Only

Formula: 164.244 g/mol
MW: C₁₁H₁₆O
Storage Temperature: Ambient
CAS Number: 1262631-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR