2-Bromo-5-(difluoromethoxy)-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

37558-5G 37558-25G 37558-1G 37558-500MG
77632-996EA 683 USD
77632-996 77659-030 77642-458 77650-464
2-Bromo-5-(difluoromethoxy)-1,3-dimethylbenzene ≥95%
2-Bromo-5-(difluoromethoxy)-1,3-dimethylbenzene

Precaución: For Research Use Only

Formula: 251.068 g/mol
MW: C₉H₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD29066628
CAS Number: 1357627-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR