α-(3,3-Diethoxy-1-propyn-1-yl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

375191-5G 375191-25G 375191-1G 375191-500MG
77628-914EA 1484.23 USD
77628-914 77635-728 77642-456 77650-460
α-(3,3-Diethoxy-1-propyn-1-yl)benzenemethanol ≥97%
α-(3,3-Diethoxy-1-propyn-1-yl)benzenemethanol

Precaución: For Research Use Only

Formula: 234.29 g/mol
MW: C₁₄H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD35196782
CAS Number: 53366-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR