1-Cyclopropyl-4,4-diethoxy-2-butyn-1-one ≥95%

Supplier: AOBChem USA

Warning

375163-5G 375163-1G 375163-500MG 375163-25G
77628-910EA 1484.23 USD
77628-910 77638-444 77652-732 77655-474
1-Cyclopropyl-4,4-diethoxy-2-butyn-1-one ≥95%
1-Cyclopropyl-4,4-diethoxy-2-butyn-1-one

Precaución: For Research Use Only

Formula: 196.24 g/mol
MW: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD20484922
CAS Number: 863672-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR