1-Bromo-4-chloro-2-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

37508-5G 37508-1G 37508-500MG
77628-908EA 1484.23 USD
77628-908 77638-442 77650-454
1-Bromo-4-chloro-2-isopropylbenzene ≥95%
1-Bromo-4-chloro-2-isopropylbenzene

Precaución: For Research Use Only

Formula: 233.533 g/mol
MW: C₉H₁₀BrCl
Storage Temperature: Ambient
MDL Number: MFCD28356422
CAS Number: 1369876-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR