2-Isopropoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

37495-25G 37495-5G
77659-028EA 160 USD
77659-028 77936-787
2-Isopropoxynaphthalene ≥97%
2-Isopropoxynaphthalene

Precaución: For Research Use Only

Formula: C₁₃H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD23114629
CAS Number: 15052-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR