4,4-Diethoxy-1-phenyl-2-butyn-1-one ≥95%

Supplier: AOBChem USA

Warning

374926-5G 374926-25G 374926-1G 374926-500MG
77628-906EA 1484.23 USD
77628-906 77635-724 77638-438 77646-474
4,4-Diethoxy-1-phenyl-2-butyn-1-one ≥95%
4,4-Diethoxy-1-phenyl-2-butyn-1-one

Precaución: For Research Use Only

Formula: 232.28 g/mol
MW: C₁₄H₁₆O₃
Storage Temperature: Ambient
CAS Number: 53366-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR