α-(3,3-Diethoxy-1-propyn-1-yl)cyclopropanemethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
374886-5G
374886-25G
374886-1G
374886-500MG
77628-904EA
1484.23
USD
77628-904
77635-722
77638-436
77646-472
α-(3,3-Diethoxy-1-propyn-1-yl)cyclopropanemethanol ≥95%
α-(3,3-Diethoxy-1-propyn-1-yl)cyclopropanemethanol
Precaución: For Research Use Only
Formula:
198.26 g/mol MW: C₁₁H₁₈O₃ Storage Temperature: Ambient |
CAS Number:
1539295-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...