N-(2-Fluorophenyl)pivalamide ≥95%

Supplier: AOBChem USA

Warning

37341-5G 37341-1G 37341-25G 37341-500MG
77632-992EA 1005 USD
77632-992 77638-434 77655-468 77650-448
N-(2-Fluorophenyl)pivalamide ≥95%
N-(2-Fluorophenyl)pivalamide

Precaución: For Research Use Only

Formula: 195.233 g/mol
MW: C₁₁H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD00995787
CAS Number: 88288-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR