5-Bromo-2,4-difluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37331-5G 37331-25G 37331-1G 37331-500MG
77634-942EA 333 USD
77634-942 77659-026 77642-450 77646-468
5-Bromo-2,4-difluoro-3-methylbenzoic acid ≥97%
5-Bromo-2,4-difluoro-3-methylbenzoic acid

Precaución: For Research Use Only

Formula: 251.025 g/mol
MW: C₈H₅BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32632199
CAS Number: 2404734-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR