4-Chloro-2-iodo-6-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

37239-5G 37239-25G 37239-1G 37239-500MG
77628-902EA 1484.23 USD
77628-902 77635-720 77638-432 77646-466
4-Chloro-2-iodo-6-(trifluoromethyl)aniline ≥97%
4-Chloro-2-iodo-6-(trifluoromethyl)aniline

Precaución: For Research Use Only

Formula: 321.466 g/mol
MW: C₇H₄ClF₃IN
Storage Temperature: Ambient
MDL Number: MFCD28010450
CAS Number: 1065102-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR