2-(2,3-Difluoro-4-methylphenyl)benzo[d]thiazole ≥95%

Supplier: AOBChem USA

Warning

37218-5G 37218-1G 37218-500MG
77628-900EA 1484.23 USD
77628-900 77642-444 77646-462
2-(2,3-Difluoro-4-methylphenyl)benzo[d]thiazole ≥95%
2-(2,3-Difluoro-4-methylphenyl)benzo[d]thiazole

Precaución: For Research Use Only

Formula: 261.29 g/mol
MW: C₁₄H₉F₂NS
Storage Temperature: Ambient
MDL Number: MFCD32632165
CAS Number: 2379321-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR