(6-Bromo-2-chloro-3-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

371889-5G 371889-1G 371889-25G 371889-500MG
77628-896EA 1484.23 USD
77628-896 77638-428 77659-014 77646-460
(6-Bromo-2-chloro-3-methylphenyl)(cyclopropyl)methanol ≥95%
(6-Bromo-2-chloro-3-methylphenyl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD35196151
CAS Number: 3002504-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR