3-Bromoquinoline-2-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

37162-1G 37162-5G 37162-500MG
77638-426EA 549 USD
77638-426 77655-464 77646-458
3-Bromoquinoline-2-carboxylic acid ≥95%
3-Bromoquinoline-2-carboxylic acid

Precaución: For Research Use Only

Formula: 252.064 g/mol
MW: C₁₀H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD26050797
CAS Number: 1564924-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR