4'-Chlorobiphenyl-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Chloro[1,1'-biphenyl]-4-carboxylic acid, 4'-Chloro-[1,1'-biphenyl]-4-carboxylic acid

37098-5G 37098-25G
77634-940EA 61 USD
77634-940 77655-462
4'-Chlorobiphenyl-4-carboxylic acid ≥97%
4'-Chlorobiphenyl-4-carboxylic acid

Precaución: For Research Use Only

Formula: C₁₃H₉ClO₂
MW: 232.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424592
CAS Number: 5748-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR