Thiophene-3-ol, tetrahydro-3-phenyl-, 1,1-dioxide ≥97%

Supplier: AOBChem USA

Warning

370698-5G 370698-1G 370698-500MG
77628-894EA 1484.23 USD
77628-894 77638-424 77650-436
Thiophene-3-ol, tetrahydro-3-phenyl-, 1,1-dioxide ≥97%
Thiophene-3-ol, tetrahydro-3-phenyl-, 1,1-dioxide

Precaución: For Research Use Only

Formula: 212.27 g/mol
MW: C₁₀H₁₂O₃S
Storage Temperature: Ambient
CAS Number: 1341435-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR