Tetrahydro-3-phenylthiophene-3-ol ≥95%

Supplier: AOBChem USA

Warning

370479-5G 370479-1G 370479-500MG
77628-892EA 1484.23 USD
77628-892 77638-422 77646-456
Tetrahydro-3-phenylthiophene-3-ol ≥95%
Tetrahydro-3-phenylthiophene-3-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD23798718
CAS Number: 91060-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR