3-(3-Methyl-5-(methylthio)phenyl)tetrahydrofuran-3-ol ≥95%

Supplier: AOBChem USA

Warning

370095-5G 370095-1G 370095-25G 370095-500MG
77628-888EA 1484.23 USD
77628-888 77638-418 77659-010 77646-454
3-(3-Methyl-5-(methylthio)phenyl)tetrahydrofuran-3-ol ≥95%
3-(3-Methyl-5-(methylthio)phenyl)tetrahydrofuran-3-ol

Precaución: For Research Use Only

Formula: 224.319 g/mol
MW: C₁₂H₁₆O₂S
Storage Temperature: Ambient
MDL Number: MFCD35195629
CAS Number: 3002489-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR