(3-Methyl-5-(methylthio)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

370091-5G 370091-25G 370091-1G 370091-500MG
77632-984EA 1441 USD
77632-984 77655-460 77642-438 77646-452
(3-Methyl-5-(methylthio)phenyl)(phenyl)methanone ≥95%
(3-Methyl-5-(methylthio)phenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₅H₁₄OS
MW: 242.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD35195625
CAS Number: 3002502-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR