3-(3-Methyl-5-(methylthio)phenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

370081-5G 370081-1G 370081-25G 370081-500MG
77628-884EA 1484.23 USD
77628-884 77638-414 77659-008 77650-432
3-(3-Methyl-5-(methylthio)phenyl)oxetan-3-ol ≥95%
3-(3-Methyl-5-(methylthio)phenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD35195615
CAS Number: 3002464-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR