1-(3-Methyl-5-(methylthio)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

370071-5G 370071-25G 370071-1G 370071-500MG
77628-882EA 1484.23 USD
77628-882 77655-458 77642-436 77646-448
1-(3-Methyl-5-(methylthio)phenyl)propan-1-one ≥95%
1-(3-Methyl-5-(methylthio)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: 194.293 g/mol
MW: C₁₁H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD35195605
CAS Number: 3002520-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR