1-(3-Methyl-5-(methylthio)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

370062-5G 370062-500MG 370062-25G 370062-1G
77628-880EA 1484.23 USD
77628-880 77652-730 77659-006 77642-434
1-(3-Methyl-5-(methylthio)phenyl)cyclobutanol ≥95%
1-(3-Methyl-5-(methylthio)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD35195596
CAS Number: 3002520-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR