(3-Chloro-5-(trifluoromethyl)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369860-5G 369860-25G 369860-1G 369860-500MG
77632-982EA 1441 USD
77632-982 77659-004 77642-432 77646-446
(3-Chloro-5-(trifluoromethyl)phenyl)(phenyl)methanone ≥95%
(3-Chloro-5-(trifluoromethyl)phenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₄H₈ClF₃O
MW: 284.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD35195412
CAS Number: 3002444-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR