(3,5-Bis(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

369796-500MG 369796-5G 369796-1G
77652-726EA 379 USD
77652-726 77655-454 77642-426
(3,5-Bis(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%
(3,5-Bis(trifluoromethyl)phenyl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₁₀F₆O
Storage Temperature: Ambient
MDL Number: MFCD25643637
CAS Number: 1512094-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR