2,2,2-Trifluoro-1-(2-methyl-5-(trifluoromethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

369730-5G 369730-1G 369730-25G 369730-500MG
77628-874EA 1648 USD
77628-874 77638-410 77655-450 77650-424
2,2,2-Trifluoro-1-(2-methyl-5-(trifluoromethyl)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-methyl-5-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 256.144 g/mol
MW: C₁₀H₆F₆O
Storage Temperature: Ambient
MDL Number: MFCD35195306
CAS Number: 2155084-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR