6-Bromo-3-chloro-2-fluoroiodobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-chloro-3-fluoro-2-iodobenzene

369726-5G 369726-25G 369726-1G 369726-500MG
77632-978EA 710 USD
77632-978 77658-998 77642-420 77646-442
6-Bromo-3-chloro-2-fluoroiodobenzene ≥95%
6-Bromo-3-chloro-2-fluoroiodobenzene

Precaución: For Research Use Only

Formula: C₆H₂BrClFI
MW: 335.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103322
CAS Number: 1820674-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR