Cyclopentyl(2-fluoro-4-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369455-5G 369455-1G 369455-500MG
77628-872EA 1484.23 USD
77628-872 77642-418 77646-440
Cyclopentyl(2-fluoro-4-methylphenyl)methanone ≥95%
Cyclopentyl(2-fluoro-4-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 206.26 g/mol
MW: C₁₃H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD28105847
CAS Number: 1602803-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR