Cyclobutyl[4-methoxy-2-(trifluoromethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

369330-5G 369330-1G 369330-500MG
77658-992EA 2590 USD
77658-992 77642-416 77646-436
Cyclobutyl[4-methoxy-2-(trifluoromethyl)phenyl]methanone ≥95%
Cyclobutyl[4-methoxy-2-(trifluoromethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD35195199
CAS Number: 2353794-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR