Cyclobutyl(4-methoxy-3,5-dimethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369276-5G 369276-1G 369276-500MG
77658-990EA 2001 USD
77658-990 77642-414 77646-434
Cyclobutyl(4-methoxy-3,5-dimethylphenyl)methanone ≥95%
Cyclobutyl(4-methoxy-3,5-dimethylphenyl)methanone

Precaución: For Research Use Only

Formula: 218.29 g/mol
MW: C₁₄H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD24363276
CAS Number: 1520906-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR