Cyclobutyl(3-methoxy-4-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369269-500MG 369269-5G 369269-1G
77652-724EA 379 USD
77652-724 77655-448 77642-412
Cyclobutyl(3-methoxy-4-methylphenyl)methanone ≥95%
Cyclobutyl(3-methoxy-4-methylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD35195181
CAS Number: 1593787-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR