Cyclobutyl[3-(1,1-dimethylethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

369206-1G 369206-5G 369206-500MG
77638-402EA 560 USD
77638-402 77658-984 77646-432
Cyclobutyl[3-(1,1-dimethylethyl)phenyl]methanone ≥95%
Cyclobutyl[3-(1,1-dimethylethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: C₁₅H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD28406643
CAS Number: 1995396-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR