Cyclobutyl(4-fluoro-2-methoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369199-500MG 369199-5G 369199-1G
77652-722EA 379 USD
77652-722 77658-982 77642-408
Cyclobutyl(4-fluoro-2-methoxyphenyl)methanone ≥95%
Cyclobutyl(4-fluoro-2-methoxyphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD28408257
CAS Number: 2091915-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR