Cyclobutyl(2,3-difluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369181-5G 369181-500MG 369181-25G 369181-1G
77632-972EA 1386 USD
77632-972 77652-720 77658-980 77642-406
Cyclobutyl(2,3-difluorophenyl)methanone ≥95%
Cyclobutyl(2,3-difluorophenyl)methanone

Precaución: For Research Use Only

Formula: 196.19 g/mol
MW: C₁₁H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD21117565
CAS Number: 1536965-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR