Cyclobutyl[4-(1,1-dimethylethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

369137-5G 369137-1G 369137-500MG
77658-978EA 2001 USD
77658-978 77646-420 77646-422
Cyclobutyl[4-(1,1-dimethylethyl)phenyl]methanone ≥95%
Cyclobutyl[4-(1,1-dimethylethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: C₁₅H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD11210409
CAS Number: 1082598-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR