Cyclobutyl(2,6-difluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369127-5G 369127-1G 369127-500MG
77628-866EA 1484.23 USD
77628-866 77638-394 77652-718
Cyclobutyl(2,6-difluorophenyl)methanone ≥95%
Cyclobutyl(2,6-difluorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₁H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD21117566
CAS Number: 1541020-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR