Cyclopropyl[2-methyl-5-(trifluoromethyl)phenyl]methanone ≥95%
Supplier: AOBChem USA
|
Warning
|
369094-5G
369094-25G
369094-1G
369094-500MG
77628-864EA
1484.23
USD
77628-864
77635-716
77642-404
77650-416
Cyclopropyl[2-methyl-5-(trifluoromethyl)phenyl]methanone ≥95%
Cyclopropyl[2-methyl-5-(trifluoromethyl)phenyl]methanone
Precaución: For Research Use Only
Formula:
C₁₂H₁₁F₃O Storage Temperature: Ambient |
MDL Number:
MFCD34147458 CAS Number: 2828726-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...