α-Cyclopropyl-2-methyl-5-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

369043-5G 369043-25G 369043-1G 369043-500MG
77628-860EA 1484.23 USD
77628-860 77635-712 77638-390 77646-416
α-Cyclopropyl-2-methyl-5-(trifluoromethyl)benzenemethanol ≥95%
α-Cyclopropyl-2-methyl-5-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 230.23 g/mol
MW: C₁₂H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD34148404
CAS Number: 2840848-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR