Ethyl 2-ethoxy-4-methoxy-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

368980-500MG 368980-5G 368980-1G
77652-712EA 462 USD
77652-712 77658-974 77642-398
Ethyl 2-ethoxy-4-methoxy-α-oxobenzeneacetate ≥95%
Ethyl 2-ethoxy-4-methoxy-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 252.26 g/mol
MW: C₁₃H₁₆O₅
Storage Temperature: Ambient
MDL Number: MFCD35195122
CAS Number: 1891763-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR