1-(2-Ethoxy-4-methoxyphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

368963-5G 368963-25G 368963-1G 368963-500MG
77628-858EA 1484.23 USD
77628-858 77658-972 77642-396 77646-414
1-(2-Ethoxy-4-methoxyphenyl)-1-propanone ≥95%
1-(2-Ethoxy-4-methoxyphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 208.25 g/mol
MW: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD13967175
CAS Number: 1114598-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR